KEY001153493 Key Organics / BIONET

ethyl 2-(2,4,5-trioxoimidazolidin-1-yl)acetate

  • MDL number MFCD00665817
  • Cas Number : 89694-35-9
  • Molecular Weight : 200.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 162
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89944

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8M-728_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart