KEY001153513 Key Organics / BIONET

5-[(1E)-3-oxo-3-phenylprop-1-en-1-yl]furan-2-carbaldehyde

  • MDL number MFCD01315261
  • Cas Number : 6575-75-3
  • Molecular Weight : 226.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105 - 107
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89968

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8M-909_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart