KEY001153556 Key Organics / BIONET

methyl 4-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzoate

  • MDL number MFCD01568814
  • Cas Number : 339105-21-4
  • Molecular Weight : 283.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108 - 109
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90011

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8N-050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart