KEY001153570 Key Organics / BIONET

1-[3-(pyrimidin-2-yloxy)phenyl]ethan-1-one

  • MDL number MFCD01568700
  • Cas Number : 339105-37-2
  • Molecular Weight : 214.224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84 - 86
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90025

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8N-068_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart