KEY001153601 Key Organics / BIONET

3-{[(3-methylphenyl)methyl]sulfanyl}-1H-1,2,4-triazole

  • MDL number MFCD01568454
  • Cas Number : 307545-27-3
  • Molecular Weight : 205.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54.6 - 55.8
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90059

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8N-367S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart