KEY001153737 Key Organics / BIONET

2-(4-methoxyphenoxy)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD01871705
  • Cas Number : 478081-25-3
  • Molecular Weight : 235.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil / low melt solid
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90200

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8P-010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart