KEY001153745 Key Organics / BIONET

2-chloro-5-[(1H-pyrazol-1-yl)methyl]-1,3-thiazole

  • MDL number MFCD01871711
  • Cas Number : 453557-78-3
  • Molecular Weight : 199.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50 - 52
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Low Melting Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90209

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/8P-025_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart