KEY001153752 Key Organics / BIONET

1,1,1-trifluoro-3-({[2-(trifluoromethyl)phenyl]methyl}amino)propan-2-ol

  • MDL number MFCD01871716
  • Cas Number : 453557-79-4
  • Molecular Weight : 287.205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79 - 81
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90217

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8P-040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart