KEY001154189 Key Organics / BIONET

methyl 1-chloroisoquinoline-4-carboxylate

  • MDL number MFCD00140353
  • Cas Number : 37497-86-2
  • Molecular Weight : 8/8/1900 15:21:36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91432

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9B-078_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart