KEY001154190 Key Organics / BIONET

2-cyano-N-(pyridin-3-yl)acetamide

  • MDL number MFCD00127642
  • Cas Number : 219618-13-0
  • Molecular Weight : 161.164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 120
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91433

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9B-079_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart