KEY001154251 Key Organics / BIONET

3-phenoxybutan-2-one

  • MDL number MFCD00243672
  • Cas Number : 6437-85-0
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 46@0.1mm Hg
Iupac Name 3-phenoxy-2-butanone
Inchi Code 1S/C10H12O2/c1-8(11)9(2)12-10-6-4-3-5-7-10/h3-7,9H,1-2H3
Inchi Key GYXROYABIGAUKY-UHFFFAOYSA-N

Description

Melting Point 46@0.1mm Hg
Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91498

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9C-042_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart