KEY001154256 Key Organics / BIONET

3-[3-(trifluoromethyl)phenoxy]butan-2-one

  • MDL number MFCD00243674
  • Cas Number : 338975-95-4
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 68@0.2mm Hg
Iupac Name 3-[3-(trifluoromethyl)phenoxy]-2-butanone
Inchi Code 1S/C11H11F3O2/c1-7(15)8(2)16-10-5-3-4-9(6-10)11(12,13)14/h3-6,8H,1-2H3
Inchi Key RUTXCSFBVGGQFH-UHFFFAOYSA-N

Description

Melting Point 68@0.2mm Hg
Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91503

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9C-053_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart