KEY001154274 Key Organics / BIONET

5-(1-phenoxyethyl)-1H-pyrazole

  • MDL number MFCD00139123
  • Cas Number : 321998-96-3
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109 - 111
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91522

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9C-082_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart