KEY001154309 Key Organics / BIONET

2,3-diamino-5-bromo-6-(trifluoromethyl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD00214813
  • Cas Number : 95095-46-8
  • Molecular Weight : 273.013

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226 - 228
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91557

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9D-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart