KEY001154313 Key Organics / BIONET

4-(4-bromophenyl)-2-oxo-2,5-dihydrofuran-3-carbonitrile

  • MDL number MFCD00139239
  • Cas Number : 7721-24-6
  • Molecular Weight : 264.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 215 - 217
Iupac Name 4-(4-bromophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile
Inchi Code 1S/C11H6BrNO2/c12-8-3-1-7(2-4-8)10-6-15-11(14)9(10)5-13/h1-4H,6H2
Inchi Key PPOYNWDABZQQRK-UHFFFAOYSA-N

Description

Melting Point 215 - 217
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91561

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9D-038_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart