KEY001154317 Key Organics / BIONET

4-(4-methoxyphenyl)-2-oxo-2,5-dihydrofuran-3-carbonitrile

  • MDL number MFCD00139243
  • Cas Number : 50691-07-1
  • Molecular Weight : 215.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 189 - 191
Iupac Name 4-(4-methoxyphenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile
Inchi Code 1S/C12H9NO3/c1-15-9-4-2-8(3-5-9)11-7-16-12(14)10(11)6-13/h2-5H,7H2,1H3
Inchi Key HOOKJRWGDDCCDO-UHFFFAOYSA-N

Description

Melting Point 189 - 191
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91565

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9D-045_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart