KEY001154471 Key Organics / BIONET

5-chloro-2-oxo-1-{[4-(trifluoromethyl)phenyl]methyl}-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00243767
  • Cas Number : 338977-66-5
  • Molecular Weight : 331.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197 - 199
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91724

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9E-368S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart