KEY001154620 Key Organics / BIONET

ethyl 2-{4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}acetate

  • MDL number MFCD00141695
  • Cas Number : 338979-08-1
  • Molecular Weight : 351.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70 - 72
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91888

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9F-910_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart