KEY001154783 Key Organics / BIONET

4-hydroxy-1-(4-methylbenzenesulfonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00215527
  • Cas Number : 20275-18-7
  • Molecular Weight : 285.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 157 - 159
Iupac Name 4-hydroxy-1-[(4-methylphenyl)sulfonyl]proline
Inchi Code 1S/C12H15NO5S/c1-8-2-4-10(5-3-8)19(17,18)13-7-9(14)6-11(13)12(15)16/h2-5,9,11,14H,6-7H2,1H3,(H,15,16)
Inchi Key UAFPTDLCLKLJKA-UHFFFAOYSA-N

Description

Melting Point 157 - 159
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92104

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9G-933_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart