KEY001154794 Key Organics / BIONET

methyl 4-hydroxy-1-(4-methylbenzenesulfonyl)pyrrolidine-2-carboxylate

  • MDL number MFCD00202789
  • Cas Number : 16257-57-1
  • Molecular Weight : 299.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97 - 99
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92121

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9G-953_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart