KEY001154923 Key Organics / BIONET

1-(4-fluorophenyl)-2-methanesulfonylethan-1-one

  • MDL number MFCD00202594
  • Cas Number : 82652-17-3
  • Molecular Weight : 216.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111 - 113
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92261

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9H-392S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart