KEY001155102 Key Organics / BIONET

2-bromo-1-(3-chlorothiophen-2-yl)ethan-1-one

  • MDL number MFCD00793892
  • Cas Number : 256486-71-2
  • Molecular Weight : 239.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 59 - 61
Iupac Name 2-bromo-1-(3-chloro-2-thienyl)ethanone
Inchi Code 1S/C6H4BrClOS/c7-3-5(9)6-4(8)1-2-10-6/h1-2H,3H2
Inchi Key HGGWBQPMDLULMR-UHFFFAOYSA-N

Description

Melting Point 59 - 61
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92487

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9J-909_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart