KEY001155194 Key Organics / BIONET

2-(trifluoromethyl)-3,4-dihydroquinazolin-4-one

  • MDL number MFCD00793796
  • Cas Number : 26059-81-4
  • Molecular Weight : 214.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248 - 251
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92614

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9L-021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart