KEY001155285 Key Organics / BIONET

2-amino-5-oxo-4-(thiophen-2-yl)-5,6,7,8-tetrahydro-4H-chromene-3-carbonitrile

  • MDL number MFCD00096530
  • Cas Number : 312510-11-5
  • Molecular Weight : 272.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92744

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9L-702_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart