KEY001155291 Key Organics / BIONET

4-[3-(furan-2-yl)-1H-pyrazol-5-yl]piperidine

  • MDL number MFCD00665128
  • Cas Number : 111897-11-1
  • Molecular Weight : 217.272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182 - 184
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92750

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9L-711_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart