KEY001155293 Key Organics / BIONET

N-[3,5-bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide

  • MDL number MFCD00793437
  • Cas Number : 478261-82-4
  • Molecular Weight : 347.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 119 - 121
Iupac Name N-[3,5-bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide
Inchi Code 1S/C13H12ClF6NO/c1-11(2,6-14)10(22)21-9-4-7(12(15,16)17)3-8(5-9)13(18,19)20/h3-5H,6H2,1-2H3,(H,21,22)
Inchi Key YFRQAWZFVGSRLL-UHFFFAOYSA-N

Description

Melting Point 119 - 121
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92752

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9L-714_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart