KEY001155301 Key Organics / BIONET

3-chloro-2,2-dimethyl-N-(1,3-thiazol-2-yl)propanamide

  • MDL number MFCD00793441
  • Cas Number : 454473-70-2
  • Molecular Weight : 218.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74 - 76
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92760

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9L-722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart