KEY001155339 Key Organics / BIONET

N-(5-iodopyridin-2-yl)acetamide

  • MDL number MFCD01315262
  • Cas Number : 66131-78-0
  • Molecular Weight : 262.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 156 - 158
Iupac Name N-(5-iodo-2-pyridinyl)acetamide
Inchi Code 1S/C7H7IN2O/c1-5(11)10-7-3-2-6(8)4-9-7/h2-4H,1H3,(H,9,10,11)
Inchi Key KUSIBMGCSKLUCE-UHFFFAOYSA-N

Description

Melting Point 156 - 158
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92798

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9L-901_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart