KEY001155392 Key Organics / BIONET

4-(dimethylamino)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD01314755
  • Cas Number : 62321-91-9
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 282 - 284
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92851

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9M-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart