KEY001155415 Key Organics / BIONET

ethyl 3-amino-4-(dimethylamino)thieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD01105700
  • Cas Number : 147992-86-7
  • Molecular Weight : 265.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 98
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92872

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9M-048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart