KEY001155545 Key Organics / BIONET

1-(furan-3-yl)ethan-1-one

  • MDL number MFCD01568491
  • Cas Number : 14313-09-8
  • Molecular Weight : 110.112

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47
Purity 97
Storage temp. Store under N2 at -18
Physical Form White to Dark Yellow Solid
Shipping Temperature Store under N2 at -18
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93014

Safety Information

signalword Warning
pcodes P411,P422, P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9M-905_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart