KEY001155912 Key Organics / BIONET

ethyl 1-(2-chloro-2,2-difluoroacetyl)piperidine-4-carboxylate

  • MDL number MFCD02082447
  • Cas Number : 454473-81-5
  • Molecular Weight : 269.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp Oil
Iupac Name ethyl 1-[chloro(difluoro)acetyl]-4-piperidinecarboxylate
Inchi Code 1S/C10H14ClF2NO3/c1-2-17-8(15)7-3-5-14(6-4-7)9(16)10(11,12)13/h7H,2-6H2,1H3
Inchi Key KSIIHIZFWNXJSH-UHFFFAOYSA-N

Description

Melting Point Oil
Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93439

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9P-906_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart