KEY001155941 Key Organics / BIONET

1-(7-bromo-5-methoxy-1-benzofuran-2-yl)ethan-1-one

  • MDL number MFCD02082020
  • Cas Number : 454473-82-6
  • Molecular Weight : 269.094

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123 - 125
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93474

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9P-962_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart