KEY001156028 Key Organics / BIONET

3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)-3-(thiophen-3-yl)propanoic acid

  • MDL number MFCD02571531
  • Cas Number : 478249-84-2
  • Molecular Weight : 287.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166 - 168
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93694

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0616_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart