KEY003445496 Key Organics / BIONET

ethyl 2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate

  • MDL number MFCD00139510
  • Cas Number : 81448-48-8
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 79 - 82
Iupac Name ethyl 2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate
Inchi Code 1S/C12H14N2O2/c1-4-16-12(15)11-9(3)13-10-7-8(2)5-6-14(10)11/h5-7H,4H2,1-3H3
Inchi Key CQNUHMHESISBRX-UHFFFAOYSA-N

Description

Melting Point 79 - 82
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/245

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/10D-547_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart