KEY003445497 Key Organics / BIONET

ethyl 2,8-dimethylimidazo[1,2-a]pyridine-3-carboxylate

  • MDL number MFCD00139511
  • Cas Number : 241146-66-7
  • Molecular Weight : 218.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79 - 82
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/246

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10D-548_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart