KEY003445806 Key Organics / BIONET

6-(2,5-dimethoxyphenyl)-2-oxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid

  • MDL number MFCD02571865
  • Cas Number : 477848-85-4
  • Molecular Weight : 278.264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243 - 245DEC
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2840

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0315_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart