KEY003445817 Key Organics / BIONET

methyl 4-cyclohexanecarbonyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD02571975
  • Cas Number : 477848-90-1
  • Molecular Weight : 235.283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168 - 170
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2854

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0336_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart