KEY003445832 Key Organics / BIONET

4-cyclohexanecarbonyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD02571980
  • Cas Number : 477848-96-7
  • Molecular Weight : 221.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220 - 222
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2874

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0361_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart