KEY003446905 Key Organics / BIONET

ethyl 2-(4-methoxyphenyl)indolizine-1-carboxylate

  • MDL number MFCD02082817
  • Cas Number : 477862-79-6
  • Molecular Weight : 295.338

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 162
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9395

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-126_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart