KEY003446906 Key Organics / BIONET

ethyl pyrrolo[1,2-c]pyrimidine-3-carboxylate

  • MDL number MFCD02082826
  • Cas Number : 107407-80-7
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 69 - 70
Iupac Name ethyl pyrrolo[1,2-c]pyrimidine-3-carboxylate
Inchi Code 1S/C10H10N2O2/c1-2-14-10(13)9-6-8-4-3-5-12(8)7-11-9/h3-7H,2H2,1H3
Inchi Key RUBOVOPFBPEEJU-UHFFFAOYSA-N

Description

Melting Point 69 - 70
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9416

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-153_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart