KEY003446995 Key Organics / BIONET

1-(2H-1,3-benzodioxol-5-yl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD03011790
  • Cas Number : 383147-55-5
  • Molecular Weight : 215.208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75 - 78
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9681

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0627_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart