KEY003447235 Key Organics / BIONET

7-[1-(2,6-dichlorophenoxy)ethyl]-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

  • MDL number MFCD00243624
  • Cas Number : 477865-30-8
  • Molecular Weight : 324.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255 - 257
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64452

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-046_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart