KEY003447473 Key Organics / BIONET

2-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carbohydrazide

  • MDL number MFCD03305519
  • Cas Number : 955964-92-8
  • Molecular Weight : 291.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206 - 208
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65915

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0899_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart