KEY003447633 Key Organics / BIONET

ethyl 2-(2-chlorophenyl)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD01935973
  • Cas Number : 338982-17-5
  • Molecular Weight : 281.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103 - 105
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67145

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2G-392S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart