KEY003447647 Key Organics / BIONET

ethyl 2-(3,4-dimethoxyphenyl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD00665469
  • Cas Number : 338982-18-6
  • Molecular Weight : 293.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 99
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67167

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2G-425S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart