KEY003447942 Key Organics / BIONET

2-(3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carbonyl)benzoic acid

  • MDL number MFCD03012223
  • Cas Number : 26513-80-4
  • Molecular Weight : 297.266

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 173
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69377

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0210_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart