KEY003448257 Key Organics / BIONET

[1,2,4]triazolo[1,5-a]pyridin-8-amine

  • MDL number MFCD01316370
  • Cas Number : 31052-95-6
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 103 - 106
Iupac Name [1,2,4]triazolo[1,5-a]pyridin-8-amine
Inchi Code 1S/C6H6N4/c7-5-2-1-3-10-6(5)8-4-9-10/h1-4H,7H2
Inchi Key LJPJXPHHXBKTKJ-UHFFFAOYSA-N

Description

Melting Point 103 - 106
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70548

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3F-375S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart