KEY003448523 Key Organics / BIONET

ethyl 7-methylimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD02187526
  • Cas Number : 70705-33-8
  • Molecular Weight : 204.229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 129
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72752

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0655_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart