KEY003448524 Key Organics / BIONET

ethyl imidazo[1,2-a]pyrimidine-2-carboxylate

  • MDL number MFCD02186164
  • Cas Number : 64951-06-0
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 - 172
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72757

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0666_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart